Product Details

SNP ID
rs10158689
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:88938286 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGCCACTCAGTTTTTCTCTCCA[A/G]AGACAACACTGGGATTTCATTATAT
Phenotype
MIM: 610656
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KYAT3 PubMed Links

Gene Details

Gene
KYAT3
Gene Name
kynurenine aminotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008661.2 Intron NP_001008661.1
NM_001008662.2 Intron NP_001008662.1

View Full Product Details