Product Details

SNP ID
rs10245796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:105025546 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATCTTCATTTAACCAATGTGCA[A/G]GATTAATAATTCTTCTTTTAAAAAT
Phenotype
MIM: 608444
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KMT2E PubMed Links

Gene Details

Gene
KMT2E
Gene Name
lysine methyltransferase 2E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018682.3 Intron NP_061152.3
NM_182931.2 Intron NP_891847.1
XM_005250493.1 Intron XP_005250550.1
XM_011516400.1 Intron XP_011514702.1
XM_017012435.1 Intron XP_016867924.1

View Full Product Details