Product Details

SNP ID
rs7553327
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:197505435 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTATAGGTTCATCATATCAAAGA[A/G]GTAATCATTTGTGACTTTCCAATAT
Phenotype
MIM: 613292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DENND1B PubMed Links

Gene Details

Gene
DENND1B
Gene Name
DENN domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195215.1 7165 UTR 3 NP_001182144.1
NM_001195216.1 7165 Intron NP_001182145.1
NM_001300858.1 7165 Intron NP_001287787.1
NM_144977.4 7165 Intron NP_659414.2
XM_005244931.2 7165 UTR 3 XP_005244988.1
XM_006711192.2 7165 UTR 3 XP_006711255.1
XM_006711193.3 7165 UTR 3 XP_006711256.1
XM_006711194.3 7165 UTR 3 XP_006711257.1
XM_011509246.2 7165 UTR 3 XP_011507548.1
XM_011509247.1 7165 UTR 3 XP_011507549.1
XM_011509248.2 7165 UTR 3 XP_011507550.1
XM_011509249.2 7165 UTR 3 XP_011507551.1
XM_011509251.2 7165 Intron XP_011507553.1
XM_017000470.1 7165 UTR 3 XP_016855959.1
XM_017000471.1 7165 UTR 3 XP_016855960.1
XM_017000472.1 7165 UTR 3 XP_016855961.1

View Full Product Details