Product Details
- SNP ID
-
rs9678342
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:230418996 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATCTATAAATCAATTTGAAGAGAA[C/T]GGACATCTTTACATTCACGAGACTT
- Phenotype
-
MIM: 604585
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SP100
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111567440] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SP100
- Gene Name
- SP100 nuclear antigen
View Full Product Details