Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_006092.2 | 3377 | Intron | NP_006083.1 | ||
XM_005249568.1 | 3377 | Intron | XP_005249625.1 | ||
XM_005249572.1 | 3377 | Intron | XP_005249629.1 | ||
XM_005249576.1 | 3377 | Intron | XP_005249633.1 | ||
XM_006715633.2 | 3377 | Intron | XP_006715696.1 | ||
XM_011515079.1 | 3377 | Intron | XP_011513381.1 | ||
XM_011515080.2 | 3377 | Intron | XP_011513382.1 | ||
XM_011515081.2 | 3377 | Intron | XP_011513383.1 | ||
XM_011515083.1 | 3377 | Intron | XP_011513385.1 | ||
XM_011515084.1 | 3377 | Intron | XP_011513386.1 | ||
XM_011515085.1 | 3377 | Intron | XP_011513387.1 | ||
XM_011515087.1 | 3377 | Intron | XP_011513389.1 | ||
XM_011515088.2 | 3377 | Intron | XP_011513390.1 | ||
XM_017011674.1 | 3377 | UTR 3 | XP_016867163.1 | ||
XM_017011675.1 | 3377 | Intron | XP_016867164.1 |