Product Details

SNP ID
rs9386926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:110611479 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTCCCACGGTTTGAGGGCCCTTA[C/T]GTGTGTTAAAAAGGCTTTGTTCCCA
Phenotype
MIM: 614720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK19 PubMed Links

Gene Details

Gene
CDK19
Gene Name
cyclin dependent kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300960.1 4328 UTR 3 NP_001287889.1
NM_001300963.1 4328 UTR 3 NP_001287892.1
NM_001300964.1 4328 UTR 3 NP_001287893.1
NM_015076.4 4328 UTR 3 NP_055891.1
XM_005266871.3 4328 UTR 3 XP_005266928.1
XM_011535630.2 4328 UTR 3 XP_011533932.1
XM_011535631.2 4328 UTR 3 XP_011533933.1
XM_011535632.1 4328 Intron XP_011533934.1
XM_017010587.1 4328 UTR 3 XP_016866076.1
XM_017010588.1 4328 UTR 3 XP_016866077.1

View Full Product Details