Product Details

SNP ID
rs6763040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112823971 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGCTCAATACGAACTATAAGAGA[A/G]GCCACTGGGGTTATGTAAGTAGAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD200R1L PubMed Links

Gene Details

Gene
CD200R1L
Gene Name
CD200 receptor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008784.2 Intron NP_001008784.2
NM_001199215.1 Intron NP_001186144.1
XM_011512763.2 Intron XP_011511065.1
XM_017006300.1 Intron XP_016861789.1
Gene
LOC100129297
Gene Name
uncharacterized LOC100129297
There are no transcripts associated with this gene.

View Full Product Details