Product Details
- SNP ID
-
rs7542144
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:3491311 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGGCCCCAGGCTCCTGAGGCCTC[C/T]AGCTCAATGTCAAGGCACTGCCCCC
- Phenotype
-
MIM: 604266
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MEGF6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114256420] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MEGF6
- Gene Name
- multiple EGF like domains 6
View Full Product Details