Product Details

SNP ID
hCV30540344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31377425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCACTGTTACCCATGCATCGAA[A/G]GATGTGGCGTTTGCCTGCCAAAGGA
Phenotype
MIM: 607650 MIM: 615997
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEFB118 PubMed Links

Gene Details

Gene
DEFB118
Gene Name
defensin beta 118
There are no transcripts associated with this gene.

Gene
DEFB119
Gene Name
defensin beta 119
There are no transcripts associated with this gene.

View Full Product Details