Product Details

SNP ID
rs6121247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31848353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGCCCCAAGTCTATGGGCTTA[A/G]TGTGGGCAGGGTTGGGTGGGAGTAG
Phenotype
MIM: 616776 MIM: 602939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012644.2 1122 Intron NP_001012662.1
NM_001320478.1 1122 Intron NP_001307407.1
NM_001320479.1 1122 UTR 3 NP_001307408.1
NM_080611.4 1122 Intron NP_542178.2
NM_177991.2 1122 Intron NP_817130.1
XM_017027654.1 1122 UTR 3 XP_016883143.1
XM_017027655.1 1122 UTR 3 XP_016883144.1
XM_017027656.1 1122 Intron XP_016883145.1
XM_017027657.1 1122 UTR 3 XP_016883146.1
XM_017027658.1 1122 UTR 3 XP_016883147.1
XM_017027659.1 1122 UTR 3 XP_016883148.1
XM_017027660.1 1122 UTR 3 XP_016883149.1
XM_017027661.1 1122 UTR 3 XP_016883150.1
XM_017027662.1 1122 UTR 3 XP_016883151.1
Gene
FOXS1
Gene Name
forkhead box S1
There are no transcripts associated with this gene.

View Full Product Details