Product Details

SNP ID
rs8132320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46466883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCTCTGAAGCCAGAGTTAAAGC[A/G]ACTTGCAAAAATGTAAAAGAATACC
Phenotype
MIM: 607711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIP2A PubMed Links

Gene Details

Gene
DIP2A
Gene Name
disco interacting protein 2 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146115.1 Intron NP_001139587.1
NM_001146116.1 Intron NP_001139588.1
NM_015151.3 Intron NP_055966.2
NM_206889.2 Intron NP_996772.1
NM_206890.2 Intron NP_996773.1
NM_206891.2 Intron NP_996774.1
XM_011529492.2 Intron XP_011527794.1
XM_011529501.1 Intron XP_011527803.1
XM_011529502.1 Intron XP_011527804.1
XM_017028291.1 Intron XP_016883780.1
XM_017028292.1 Intron XP_016883781.1
XM_017028293.1 Intron XP_016883782.1
XM_017028294.1 Intron XP_016883783.1
XM_017028295.1 Intron XP_016883784.1
XM_017028296.1 Intron XP_016883785.1
XM_017028297.1 Intron XP_016883786.1
XM_017028298.1 Intron XP_016883787.1
XM_017028299.1 Intron XP_016883788.1
XM_017028300.1 Intron XP_016883789.1
XM_017028301.1 Intron XP_016883790.1
XM_017028302.1 Intron XP_016883791.1
XM_017028303.1 Intron XP_016883792.1
XM_017028304.1 Intron XP_016883793.1
XM_017028305.1 Intron XP_016883794.1
Gene
DIP2A-IT1
Gene Name
DIP2A intronic transcript 1
There are no transcripts associated with this gene.

View Full Product Details