Product Details

SNP ID
rs9786246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:12665911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACAGTTTTAGAAAGTAGGTATT[C/T]GTCTCAACTTCAACATTCACTTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTTY15 PubMed Links

Gene Details

Gene
TTTY15
Gene Name
testis-specific transcript, Y-linked 15 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details