Product Details

SNP ID
rs6070549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58661695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCTGGTTGTGATTGTGCGTTCA[A/G]GACTCTGAAGTGCCAGGTACTGGCC
Phenotype
MIM: 603666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STX16 PubMed Links

Gene Details

Gene
STX16
Gene Name
syntaxin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001433.2 Intron NP_001001433.1
NM_001134772.2 Intron NP_001128244.1
NM_001134773.2 Intron NP_001128245.1
NM_001204868.1 Intron NP_001191797.1
NM_003763.5 Intron NP_003754.2
Gene
STX16-NPEPL1
Gene Name
STX16-NPEPL1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details