Product Details

SNP ID
rs133309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42002602 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTCTGTCTCAAAACAAAACAAA[A/G]TAAAAAAACTCAGTCTATCTACAGT
Phenotype
MIM: 608314 MIM: 610981
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPT3 PubMed Links

Gene Details

Gene
SEPT3
Gene Name
septin 3
There are no transcripts associated with this gene.

Gene
WBP2NL
Gene Name
WBP2 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152613.2 Intron NP_689826.2

View Full Product Details