Product Details

SNP ID
rs9971902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:70374538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTAATGTGTAAAAAATTGTACTT[A/G]TGAGTTGTAATATCCTCTTCTATTC
Phenotype
MIM: 605223
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNMB4 PubMed Links

Gene Details

Gene
KCNMB4
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014505.5 Intron NP_055320.4
XM_011538188.2 Intron XP_011536490.1

View Full Product Details