Product Details

SNP ID
rs10410564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56157157 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTCCAGGGTGGTGCTCTGGGCC[C/T]GAGTGAGACTGTGCTCTTCTGACTC
Phenotype
MIM: 607874
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF444 PubMed Links

Gene Details

Gene
ZNF444
Gene Name
zinc finger protein 444
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253792.1 1945 Intron NP_001240721.1
NM_018337.3 1945 Intron NP_060807.2
XM_005259034.2 1945 Intron XP_005259091.1
XM_005259035.1 1945 Intron XP_005259092.1
XM_005259036.1 1945 Intron XP_005259093.1
XM_011527068.1 1945 Intron XP_011525370.1
XM_011527069.1 1945 Intron XP_011525371.1
XM_011527070.1 1945 UTR 5 XP_011525372.1
XM_011527071.1 1945 Intron XP_011525373.1
XM_011527073.1 1945 UTR 5 XP_011525375.1
XM_017026932.1 1945 Intron XP_016882421.1
XM_017026933.1 1945 Intron XP_016882422.1

View Full Product Details