Product Details

SNP ID
rs9471761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42104641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCTCCGGGCTGCGGCCCGAAGC[A/G]GTTGGGAGAGCTCAGGCTGCGGCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C6orf132 PubMed Links

Gene Details

Gene
C6orf132
Gene Name
chromosome 6 open reading frame 132
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164446.2 3804 Missense Mutation CGC,TGC R1091C NP_001157918.1

View Full Product Details