Product Details

SNP ID
rs10113423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39316576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTATCACTGATAATCTTTGAATT[C/T]CTCACTTGTCTAATTTCCAGAATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAM5 PubMed Links

Gene Details

Gene
ADAM5
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details