Product Details

SNP ID
rs10065172
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150848436 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCACCACAACCCTGGAGAACTAC[C/T]TGATGGAAATGCAGTTCAACCGGTA
Phenotype
MIM: 608212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRGM PubMed Links

Gene Details

Gene
IRGM
Gene Name
immunity related GTPase M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145805.1 1426 Silent Mutation CTG,TTG L105L NP_001139277.1
XM_011537641.2 1426 Silent Mutation CTG,TTG L105L XP_011535943.1

View Full Product Details