Product Details

SNP ID
rs9398428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116038898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGCACTCTCCCCACTGCCTATA[C/T]CGACTTTGCCCCAGAAGCTAGAGCC
Phenotype
MIM: 606573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRK PubMed Links

Gene Details

Gene
FRK
Gene Name
fyn related Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002031.2 Intron NP_002022.1
XM_005266880.4 Intron XP_005266937.1
XM_005266881.2 Intron XP_005266938.1
XM_005266882.4 Intron XP_005266939.1
XM_011535653.2 Intron XP_011533955.1
XM_011535654.2 Intron XP_011533956.1
XM_011535655.2 Intron XP_011533957.1
XM_011535656.2 Intron XP_011533958.1
XM_017010645.1 Intron XP_016866134.1
Gene
TPI1P3
Gene Name
triosephosphate isomerase 1 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details