Product Details

SNP ID
hCV30598327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8223731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACGTGGCCGCATGAATGTGGT[C/G]GAAGGCCTGTAGACCCGGTTTGGGG
Phenotype
MIM: 613129
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTC1 PubMed Links

Gene Details

Gene
CTC1
Gene Name
CTS telomere maintenance complex component 1
There are no transcripts associated with this gene.

Gene
LINC00324
Gene Name
long intergenic non-protein coding RNA 324
There are no transcripts associated with this gene.

View Full Product Details