Product Details

SNP ID
rs9838374
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:193598916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTTGAAAGAAGGGTGAGCTAAA[C/T]GTTGTGGAACCATGAGCTCTTTCTC
Phenotype
MIM: 605290
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OPA1 PubMed Links
Additional Information
For this assay, SNP(s) [rs111944669] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OPA1
Gene Name
OPA1, mitochondrial dynamin like GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015560.2 Intron NP_056375.2
NM_130831.2 Intron NP_570844.1
NM_130832.2 Intron NP_570845.1
NM_130833.2 Intron NP_570846.1
NM_130834.2 Intron NP_570847.2
NM_130835.2 Intron NP_570848.1
NM_130836.2 Intron NP_570849.2
NM_130837.2 Intron NP_570850.2
XM_017006500.1 Intron XP_016861989.1

View Full Product Details