Product Details

SNP ID
rs8176320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43044346 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGTGTTAACAGCACAACATTTA[C/T]AAAACGTATTTTGTACAATCAAGTC
Phenotype
MIM: 113705
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRCA1 PubMed Links

Gene Details

Gene
BRCA1
Gene Name
BRCA1, DNA repair associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007294.3 7219 UTR 3 NP_009225.1
NM_007297.3 7219 UTR 3 NP_009228.2
NM_007298.3 7219 UTR 3 NP_009229.2
NM_007299.3 7219 UTR 3 NP_009230.2
NM_007300.3 7219 UTR 3 NP_009231.2

View Full Product Details