Product Details

SNP ID
rs10028754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:5714724 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTACTGCAAGTTAAGGATCAGTT[A/G]TGGTCATGAATACCTTCTTTTCTTG
Phenotype
MIM: 604831 MIM: 607261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EVC PubMed Links

Gene Details

Gene
EVC
Gene Name
EvC ciliary complex subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306090.1 Intron NP_001293019.1
NM_001306092.1 Intron NP_001293021.1
NM_153717.2 Intron NP_714928.1
XM_006713865.3 Intron XP_006713928.1
XM_006713866.3 Intron XP_006713929.1
XM_011513419.2 Intron XP_011511721.1
XM_017007883.1 Intron XP_016863372.1
Gene
EVC2
Gene Name
EvC ciliary complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details