Product Details

SNP ID
hCV30609355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143599638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGAGGACGAGGAGGAGGCCACC[G/T]ACTATGGAGGGACCTCAGTGCCGAC
Phenotype
MIM: 130592 MIM: 616408
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EEF1D PubMed Links

Gene Details

Gene
EEF1D
Gene Name
eukaryotic translation elongation factor 1 delta
There are no transcripts associated with this gene.

Gene
PYCRL
Gene Name
pyrroline-5-carboxylate reductase-like
There are no transcripts associated with this gene.

Gene
TIGD5
Gene Name
tigger transposable element derived 5
There are no transcripts associated with this gene.

View Full Product Details