Product Details

SNP ID
rs5987189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153977113 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAATACAAAATGCCAGCTTCATAG[C/T]ATGAATGGATCCTGCTGTTCTGCAC
Phenotype
MIM: 300019 MIM: 300059
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCFC1 PubMed Links

Gene Details

Gene
HCFC1
Gene Name
host cell factor C1
There are no transcripts associated with this gene.

Gene
HCFC1-AS1
Gene Name
HCFC1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MIR3202-1
Gene Name
microRNA 3202-1
There are no transcripts associated with this gene.

Gene
MIR3202-2
Gene Name
microRNA 3202-2
There are no transcripts associated with this gene.

Gene
TMEM187
Gene Name
transmembrane protein 187
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003492.2 Intron NP_003483.1
XM_011531199.2 Intron XP_011529501.1

View Full Product Details