Product Details

SNP ID
rs6089701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62240390 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTATTAATGACAGCTCTTACCCC[A/G]GCAGACGATAATATTTAATAATGAT
Phenotype
MIM: 606731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105369209 PubMed Links

Gene Details

Gene
LOC105369209
Gene Name
putative uncharacterized protein FLJ44790
There are no transcripts associated with this gene.

Gene
OSBPL2
Gene Name
oxysterol binding protein like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278649.1 Intron NP_001265578.1
NM_014835.3 Intron NP_055650.1
NM_144498.2 Intron NP_653081.1
XM_017028163.1 Intron XP_016883652.1
XM_017028164.1 Intron XP_016883653.1
XM_017028165.1 Intron XP_016883654.1
XM_017028166.1 Intron XP_016883655.1
XM_017028167.1 Intron XP_016883656.1
XM_017028168.1 Intron XP_016883657.1
XM_017028169.1 Intron XP_016883658.1
XM_017028170.1 Intron XP_016883659.1

View Full Product Details