Product Details

SNP ID
rs6089103
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:31877116 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAGGAGTTCCTTGAGCACCAAT[A/C]CTTTGTTATATATGTTGTAAGTGTT
Phenotype
MIM: 616776
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
DUSP15 PubMed Links

Gene Details

Gene
DUSP15
Gene Name
dual specificity phosphatase 15
There are no transcripts associated with this gene.

Gene
TTLL9
Gene Name
tubulin tyrosine ligase like 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008409.2 Intron NP_001008409.1
XM_005260309.2 Intron XP_005260366.1
XM_006723723.2 Intron XP_006723786.1
XM_006723724.2 Intron XP_006723787.1
XM_011528617.1 Intron XP_011526919.1
XM_011528618.1 Intron XP_011526920.1
XM_011528619.1 Intron XP_011526921.1
XM_011528620.2 Intron XP_011526922.1
XM_011528621.1 Intron XP_011526923.1
XM_011528622.1 Intron XP_011526924.1
XM_011528623.2 Intron XP_011526925.1
XM_011528624.1 Intron XP_011526926.1
XM_011528625.1 Intron XP_011526927.1
XM_011528626.1 Intron XP_011526928.1
XM_011528627.2 Intron XP_011526929.1
XM_011528628.1 Intron XP_011526930.1
XM_011528629.1 Intron XP_011526931.1
XM_011528630.1 Intron XP_011526932.1
XM_011528631.1 Intron XP_011526933.1
XM_011528632.1 Intron XP_011526934.1
XM_011528633.1 Intron XP_011526935.1
XM_011528634.1 Intron XP_011526936.1
XM_011528635.1 Intron XP_011526937.1
XM_011528637.2 Intron XP_011526939.1
XM_011528638.1 Intron XP_011526940.1
XM_011528639.1 Intron XP_011526941.1
XM_011528640.1 Intron XP_011526942.1
XM_011528641.1 Intron XP_011526943.1
XM_011528643.2 Intron XP_011526945.1
XM_011528644.1 Intron XP_011526946.1
XM_011528646.1 Intron XP_011526948.1
XM_017027694.1 Intron XP_016883183.1
XM_017027695.1 Intron XP_016883184.1
XM_017027696.1 Intron XP_016883185.1
XM_017027697.1 Intron XP_016883186.1
XM_017027698.1 Intron XP_016883187.1
XM_017027699.1 Intron XP_016883188.1
XM_017027700.1 Intron XP_016883189.1
XM_017027701.1 Intron XP_016883190.1
XM_017027702.1 Intron XP_016883191.1
XM_017027703.1 Intron XP_016883192.1

View Full Product Details