Product Details

SNP ID
rs9831364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113288319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCATCCTTTTTCCATCAAGCTCAA[C/T]GTCAGAGAGAACTTCATGTGGAAGT
Phenotype
MIM: 608708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOC PubMed Links

Gene Details

Gene
BOC
Gene Name
BOC cell adhesion associated, oncogene regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301861.1 8298 Intron NP_001288790.1
NM_001301867.1 8298 Intron NP_001288796.1
NM_033254.3 8298 Intron NP_150279.1
XM_005247891.2 8298 Intron XP_005247948.1
XM_005247892.2 8298 Intron XP_005247949.1
XM_005247897.2 8298 Intron XP_005247954.1
XM_011513305.2 8298 Intron XP_011511607.1
XM_017007441.1 8298 Intron XP_016862930.1
XM_017007442.1 8298 Intron XP_016862931.1
XM_017007443.1 8298 Intron XP_016862932.1
XM_017007444.1 8298 Intron XP_016862933.1
XM_017007445.1 8298 Intron XP_016862934.1
XM_017007446.1 8298 Intron XP_016862935.1
XM_017007447.1 8298 Intron XP_016862936.1
XM_017007448.1 8298 Intron XP_016862937.1
XM_017007449.1 8298 Intron XP_016862938.1
XM_017007450.1 8298 Intron XP_016862939.1
XM_017007451.1 8298 Intron XP_016862940.1
XM_017007452.1 8298 Intron XP_016862941.1
XM_017007453.1 8298 Intron XP_016862942.1
XM_017007454.1 8298 Intron XP_016862943.1
XM_017007455.1 8298 Intron XP_016862944.1
Gene
CFAP44
Gene Name
cilia and flagella associated protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164496.1 8298 UTR 3 NP_001157968.1
NM_018338.3 8298 Intron NP_060808.2
XM_005247617.3 8298 UTR 3 XP_005247674.1
XM_006713696.2 8298 UTR 3 XP_006713759.1
XM_011512977.2 8298 UTR 3 XP_011511279.1
XM_011512978.2 8298 UTR 3 XP_011511280.1
XM_011512979.2 8298 UTR 3 XP_011511281.1
XM_011512980.2 8298 Intron XP_011511282.1
XM_011512981.1 8298 Intron XP_011511283.1
XM_011512982.2 8298 Intron XP_011511284.1
XM_011512983.2 8298 Intron XP_011511285.1
XM_011512984.2 8298 Intron XP_011511286.1
XM_017006842.1 8298 Intron XP_016862331.1
XM_017006843.1 8298 Intron XP_016862332.1
XM_017006844.1 8298 UTR 3 XP_016862333.1
XM_017006845.1 8298 UTR 3 XP_016862334.1
XM_017006846.1 8298 UTR 3 XP_016862335.1
XM_017006847.1 8298 UTR 3 XP_016862336.1
XM_017006848.1 8298 Intron XP_016862337.1
XM_017006849.1 8298 Intron XP_016862338.1

View Full Product Details