Product Details

SNP ID
hCV30618070
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.1:235447778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTCAGATTAAGAAAGAAATAC[A/G]CTACTGAAATGGTATGAAACTCACT
Phenotype
MIM: 610194 MIM: 604934
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
B3GALNT2 PubMed Links

Gene Details

Gene
B3GALNT2
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
There are no transcripts associated with this gene.

Gene
TBCE
Gene Name
tubulin folding cofactor E
There are no transcripts associated with this gene.

View Full Product Details