Product Details

SNP ID
rs10495306
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:231247427 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACGAGTTTAATGAAGTGTTTTA[C/G]AGACCATCTTGAAAATCCGATTATT
Phenotype
MIM: 602744
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf131 PubMed Links

Gene Details

Gene
C1orf131
Gene Name
chromosome 1 open reading frame 131
There are no transcripts associated with this gene.

Gene
GNPAT
Gene Name
glyceronephosphate O-acyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316350.1 Intron NP_001303279.1
NM_014236.3 Intron NP_055051.1
XM_005273313.4 Intron XP_005273370.1
XM_011544303.2 Intron XP_011542605.1
XM_011544304.2 Intron XP_011542606.1

View Full Product Details