Product Details

SNP ID
rs10181401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:238246073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCTGGGAGCACTGAGGCAACT[A/T]CCCTGACACTAAGAGGCGCTTAGTC
Phenotype
MIM: 610331 MIM: 603426
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HES6 PubMed Links

Gene Details

Gene
HES6
Gene Name
hes family bHLH transcription factor 6
There are no transcripts associated with this gene.

Gene
LOC101927958
Gene Name
uncharacterized LOC101927958
There are no transcripts associated with this gene.

Gene
PER2
Gene Name
period circadian clock 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022817.2 4245 UTR 3 NP_073728.1
XM_005246111.4 4245 UTR 3 XP_005246168.1
XM_006712824.3 4245 UTR 3 XP_006712887.1

View Full Product Details