Product Details

SNP ID
rs9319180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:85793394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGCATTTGTTTAAGCTCAAAT[A/G]TTCAAAGTTTTGAGCCACCCATGGC
Phenotype
MIM: 609681
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLITRK6 PubMed Links

Gene Details

Gene
SLITRK6
Gene Name
SLIT and NTRK like family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032229.2 3574 UTR 3 NP_115605.2

View Full Product Details