Product Details

SNP ID
rs9623413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41527314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATGGCATCAGGTGGGTGGTGAT[C/T]GGAGACGAGAACTACGGCGAGGGCT
Phenotype
MIM: 100850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACO2 PubMed Links

Gene Details

Gene
ACO2
Gene Name
aconitase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098.2 2002 Silent Mutation ATC,ATT I660I NP_001089.1
XM_017028812.1 2002 Silent Mutation ATC,ATT I627I XP_016884301.1
Gene
POLR3H
Gene Name
RNA polymerase III subunit H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018050.3 2002 UTR 3 NP_001018060.1
NM_001018052.3 2002 UTR 3 NP_001018062.1
NM_001282884.1 2002 UTR 3 NP_001269813.1
NM_001282885.1 2002 UTR 3 NP_001269814.1
NM_138338.4 2002 UTR 3 NP_612211.1
XM_011529993.1 2002 Intron XP_011528295.1
XM_017028641.1 2002 Intron XP_016884130.1

View Full Product Details