Product Details

SNP ID
rs7412407
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:12749993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAATTTTGGGATGTAACTCTACT[C/G]TTTATTTTTGACAGAATTTAGAAGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf158 PubMed Links

Gene Details

Gene
C1orf158
Gene Name
chromosome 1 open reading frame 158
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152290.3 Intron NP_689503.3
XM_011542434.1 Intron XP_011540736.1

View Full Product Details