Product Details

SNP ID
rs10215841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112429535 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTTGTCTTTGTTGTTCACCACA[C/T]TGCCACCTGATAAATGGCTTCTAAG
Phenotype
MIM: 603502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFRD1 PubMed Links

Gene Details

Gene
IFRD1
Gene Name
interferon related developmental regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007245.2 Intron NP_001007246.1
NM_001197079.1 Intron NP_001184008.1
NM_001197080.1 Intron NP_001184009.1
NM_001550.3 Intron NP_001541.2
XM_011516142.2 Intron XP_011514444.1
XM_011516143.2 Intron XP_011514445.1
XM_017012151.1 Intron XP_016867640.1

View Full Product Details