Product Details

SNP ID
rs1051874
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:52792883 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCAGCAGAGGGAAGCTGGAGATAAG[A/G]CTGGAGTCAAGCTCCTCGACGTAGT
Phenotype
MIM: 138359
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTA1 PubMed Links

Gene Details

Gene
GSTA1
Gene Name
glutathione S-transferase alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319059.1 732 Silent Mutation AGC,AGT S80S NP_001305988.1
NM_145740.4 732 Silent Mutation AGC,AGT S173S NP_665683.1
XM_005249034.3 732 Silent Mutation AGC,AGT S173S XP_005249091.1

View Full Product Details