Product Details

SNP ID
rs72559710
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.10:133528530 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TTCACGCTGTACGTGGGCTCGCAGC[G/A]CATGGTGGTGATGCACGGCTACAAG
Phenotype
MIM: 124040
Polymorphism
G/A, Transition substitution
Allele Nomenclature
CYP2E1*2,g.1132G>A
Literature Links
CYP2E1 PubMed Links
Additional Information
The CYP2E1 gene exhibits copy number variation. Individuals may carry extra copies of CYP2E1. CYP2E1 SNP genotyping assays run on samples having 2 or more gene copies that are homozygous for the SNP allele will cluster together, and samples having more than 2 gene copies that are heterozygous may run between the 2 copy heterozygous and homozygous clusters. For accurate CYP2E1 genotype analysis, copy number analysis must be done. For more information, refer to the PGx Experiments User Guide (Pub. # MAN0009612) Chapter 2 Copy Number Variation section.

Gene Details

Gene
CYP2E1
Gene Name
cytochrome P450 family 2 subfamily E member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000773.3 260 Missense Mutation CAC,CGC H76R NP_000764.1

View Full Product Details