Product Details

SNP ID
rs56160784
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.15:74749801 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AGCTTCTCCTGGCCTCTGCCATCTT[C/G]TGCCTGGTATTCTGGGTGCTCAAGG
Phenotype
MIM: 124060
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
CYP1A2*2,g.63C>G
Literature Links
CYP1A2 PubMed Links

Gene Details

Gene
CYP1A2
Gene Name
cytochrome P450 family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000761.4 127 Missense Mutation TTC,TTG F21L NP_000752.2

View Full Product Details