Product Details

SNP ID
rs5745078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132625317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCAGCTTGGCGCGTCCCCCAGCC[G/T]CTGCTCAGATGCCCCTCGGCAAGAC
Phenotype
MIM: 600844 MIM: 174762
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
P2RX2 PubMed Links

Gene Details

Gene
P2RX2
Gene Name
purinergic receptor P2X 2
There are no transcripts associated with this gene.

Gene
POLE
Gene Name
DNA polymerase epsilon, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006231.3 6964 Intron NP_006222.2
XM_011534795.2 6964 Silent Mutation AGG,CGG R2254R XP_011533097.1
XM_011534797.2 6964 Silent Mutation AGG,CGG R1947R XP_011533099.1
XM_011534798.1 6964 Silent Mutation AGG,CGG R1808R XP_011533100.1
XM_011534799.2 6964 Intron XP_011533101.1
XM_011534800.2 6964 Intron XP_011533102.1
XM_011534802.2 6964 Silent Mutation AGG,CGG R1250R XP_011533104.1

View Full Product Details