Product Details
- SNP ID
-
rs7598916
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:88527080 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACACCGATGTAACCATTACAGAC[C/A]GCTTAAAACATGGCACATTAACATT
- Phenotype
-
- Polymorphism
- C/A, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TEX37
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116556790] are located under a probe and SNP(s) [rs76629314] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TEX37
- Gene Name
- testis expressed 37
View Full Product Details