Product Details

SNP ID
rs13146669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:158678391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTTCTCATGATTAGACTGAGG[G/T]TAAAGATTTTTGGGGATAAATACCA
Phenotype
MIM: 616210 MIM: 231675
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C4orf46 PubMed Links

Gene Details

Gene
C4orf46
Gene Name
chromosome 4 open reading frame 46
There are no transcripts associated with this gene.

Gene
ETFDH
Gene Name
electron transfer flavoprotein dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281737.1 Intron NP_001268666.1
NM_001281738.1 Intron NP_001268667.1
NM_004453.3 Intron NP_004444.2

View Full Product Details