Product Details

SNP ID
rs11718159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48938911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCTGTCCCCACTGGGTCTTTAG[A/C]CCAAAAAAAAAAAAAAAAAAAAAAA
Phenotype
MIM: 605615
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARIH2 PubMed Links

Gene Details

Gene
ARIH2
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317333.1 Intron NP_001304262.1
NM_001317334.1 Intron NP_001304263.1
NM_006321.3 Intron NP_006312.1
XM_011533265.1 Intron XP_011531567.1
XM_011533266.1 Intron XP_011531568.1
XM_011533267.1 Intron XP_011531569.1
XM_011533268.1 Intron XP_011531570.1
XM_011533270.1 Intron XP_011531572.1
XM_011533271.1 Intron XP_011531573.1
XM_011533273.1 Intron XP_011531575.1
XM_011533275.2 Intron XP_011531577.1
XM_011533276.1 Intron XP_011531578.1
XM_017005531.1 Intron XP_016861020.1
XM_017005532.1 Intron XP_016861021.1
XM_017005533.1 Intron XP_016861022.1
XM_017005534.1 Intron XP_016861023.1
XM_017005535.1 Intron XP_016861024.1
XM_017005536.1 Intron XP_016861025.1
XM_017005537.1 Intron XP_016861026.1
XM_017005538.1 Intron XP_016861027.1
XM_017005539.1 Intron XP_016861028.1
XM_017005540.1 Intron XP_016861029.1
XM_017005541.1 Intron XP_016861030.1

View Full Product Details