Product Details

SNP ID
rs12515399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179681501 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCTGAGCCAGAGTGTGGTCTCAC[A/G]TGGAGACTAGCTTCCGCTTGCTCTC
Phenotype
MIM: 114217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CANX PubMed Links

Gene Details

Gene
CANX
Gene Name
calnexin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024649.1 330 Intron NP_001019820.1
NM_001746.3 330 Intron NP_001737.1
XM_011534664.1 330 Intron XP_011532966.1
XM_011534665.2 330 Intron XP_011532967.1
Gene
CBY3
Gene Name
chibby family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164444.1 330 Intron NP_001157916.1
XM_017009726.1 330 UTR 5 XP_016865215.1
Gene
LOC105377763
Gene Name
uncharacterized LOC105377763
There are no transcripts associated with this gene.

View Full Product Details