Product Details

SNP ID
rs13138378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76005833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACGAAGCAAGTAAGGAAGATTTA[C/G]AAAACTGTATACTAAGGGGTTAACA
Phenotype
MIM: 603086 MIM: 601704
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ART3 PubMed Links

Gene Details

Gene
ART3
Gene Name
ADP-ribosyltransferase 3
There are no transcripts associated with this gene.

Gene
CXCL9
Gene Name
C-X-C motif chemokine ligand 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002416.2 Intron NP_002407.1
Gene
LOC101928809
Gene Name
uncharacterized LOC101928809
There are no transcripts associated with this gene.

View Full Product Details