Product Details

SNP ID
rs12049195
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:156063096 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAGTATTGAGGGTTGGGTTGG[A/G]GTCCCTAGAAGCAGACGCTGAGAAG
Phenotype
MIM: 610389 MIM: 611007 MIM: 612942 MIM: 605440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAMTOR2 PubMed Links

Gene Details

Gene
LAMTOR2
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
There are no transcripts associated with this gene.

Gene
MEX3A
Gene Name
mex-3 RNA binding family member A
There are no transcripts associated with this gene.

Gene
RAB25
Gene Name
RAB25, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020387.3 Intron NP_065120.2
Gene
UBQLN4
Gene Name
ubiquilin 4
There are no transcripts associated with this gene.

View Full Product Details