Product Details

SNP ID
rs6672645
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:36459556 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGATAAGCAGGCCTTTTGTGGC[C/T]GAGCTGGTCCCTAGCCCAGGGCTTT
Phenotype
MIM: 138971 MIM: 611979 MIM: 608854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSF3R PubMed Links

Gene Details

Gene
CSF3R
Gene Name
colony stimulating factor 3 receptor
There are no transcripts associated with this gene.

Gene
MRPS15
Gene Name
mitochondrial ribosomal protein S15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031280.3 Intron NP_112570.2
Gene
OSCP1
Gene Name
organic solute carrier partner 1
There are no transcripts associated with this gene.

View Full Product Details