Product Details

SNP ID
rs12161523
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:27250739 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATATTACCATTCAATTCTTGGTT[C/T]TAGATGAGTGATCTTCACAAATGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDTC1 PubMed Links

Gene Details

Gene
WDTC1
Gene Name
WD and tetratricopeptide repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276252.1 Intron NP_001263181.1
NM_015023.4 Intron NP_055838.2
XM_011541057.1 Intron XP_011539359.1
XM_011541058.2 Intron XP_011539360.1
XM_017000730.1 Intron XP_016856219.1

View Full Product Details