Product Details

SNP ID
rs10736886
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:126910599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTAAAATATAATTCAGGGAGATC[A/G]TGAGTAGCCATTACTAGTTTCACCT
Phenotype
MIM: 601403
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DOCK1 PubMed Links

Gene Details

Gene
DOCK1
Gene Name
dedicator of cytokinesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290223.1 Intron NP_001277152.1
NM_001380.4 Intron NP_001371.1
XM_011539422.2 Intron XP_011537724.1
XM_011539424.1 Intron XP_011537726.1
XM_017015813.1 Intron XP_016871302.1
XM_017015814.1 Intron XP_016871303.1
XM_017015815.1 Intron XP_016871304.1
XM_017015816.1 Intron XP_016871305.1
XM_017015817.1 Intron XP_016871306.1

View Full Product Details