Product Details

SNP ID
rs11128117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69169199 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCATGGTATTTTGTAATAAAAAA[A/G]AGGAAACGTACATGTTGAATAATGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRMD4B PubMed Links

Gene Details

Gene
FRMD4B
Gene Name
FERM domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015123.2 6082 UTR 3 NP_055938.1
XM_005264722.1 6082 Intron XP_005264779.1
XM_005264723.1 6082 Intron XP_005264780.1
XM_017005989.1 6082 Intron XP_016861478.1
XM_017005990.1 6082 Intron XP_016861479.1
XM_017005991.1 6082 Intron XP_016861480.1
XM_017005992.1 6082 Intron XP_016861481.1
XM_017005993.1 6082 Intron XP_016861482.1
XM_017005994.1 6082 Intron XP_016861483.1
XM_017005995.1 6082 Intron XP_016861484.1
XM_017005996.1 6082 Intron XP_016861485.1
XM_017005997.1 6082 Intron XP_016861486.1

View Full Product Details